(4-Chloro-2-isopropoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

53204-500MG 53204-1G 53204-5G
77340-260EA 863.14 USD
77340-260 77352-680 77376-916
(4-Chloro-2-isopropoxyphenyl)methanol ≥97%
(4-Chloro-2-isopropoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD27931548
CAS Number: 888739-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR