4-Chloro-1-iodo-2-isopropoxybenzene ≥95%

Supplier: AOBChem USA

Warning

53200-500MG 53200-1G 53200-5G
77335-556EA 357.41 USD
77335-556 77352-674 77376-910
4-Chloro-1-iodo-2-isopropoxybenzene ≥95%
4-Chloro-1-iodo-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀ClIO
MW: 296.533 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29092605
CAS Number: 1369886-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR