4-Chloro-2-ethoxy-1-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

53199-500MG 53199-1G 53199-5G
77336-416EA 387.28 USD
77336-416 77352-672 77376-908
4-Chloro-2-ethoxy-1-iodobenzene ≥95%
4-Chloro-2-ethoxy-1-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₈ClIO
MW: 282.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20483888
CAS Number: 408326-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR