4-Chloro-2-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53157-500MG 53157-1G 53157-5G 53157-25G
77328-936EA 233.81 USD
77328-936 77352-666 77365-228 77387-298
4-Chloro-2-ethoxybenzoic acid ≥97%
4-Chloro-2-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MW: 200.619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14646682
CAS Number: 1195264-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR