2-(Benzyloxy)-4-chlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53152-500MG 53152-1G 53152-5G 53152-25G
77328-932EA 279.13 USD
77328-932 77352-660 77365-226 77387-294
2-(Benzyloxy)-4-chlorobenzoic acid ≥97%
2-(Benzyloxy)-4-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₃
MW: 262.688 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13227458
CAS Number: 57479-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR