(2-Bromo-5-chlorophenoxy)(tert-butyl)dimethylsilane ≥95%

Supplier: AOBChem USA

Warning

53147-500MG 53147-1G 53147-5G
77336-972EA 401.7 USD
77336-972 77352-658 77376-894
(2-Bromo-5-chlorophenoxy)(tert-butyl)dimethylsilane ≥95%
(2-Bromo-5-chlorophenoxy)(tert-butyl)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₂H₁₈BrClOSi
MW: 321.713 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178672
CAS Number: 174265-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR