(4-Chloro-2-methoxy-3-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

53136-500MG 53136-1G
77340-548EA 1738.64 USD
77340-548 77352-654
(4-Chloro-2-methoxy-3-methylphenyl)(methyl)sulfane ≥95%
(4-Chloro-2-methoxy-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁ClOS
MW: 202.701 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756980
CAS Number: 2708132-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR