Methyl 6-chloro-2-fluoro-3-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

53129-500MG 53129-1G 53129-5G
77336-624EA 390.37 USD
77336-624 77352-648 77376-888
Methyl 6-chloro-2-fluoro-3-(methylthio)benzoate ≥97%
Methyl 6-chloro-2-fluoro-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂S
MW: 234.675 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756979
CAS Number: 2755717-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR