6-Chloro-2-fluoro-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

53128-500MG 53128-1G 53128-5G
77328-928EA 326.51 USD
77328-928 77352-646 77376-886
6-Chloro-2-fluoro-3-(methylthio)benzoic acid ≥97%
6-Chloro-2-fluoro-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂S
MW: 220.648 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22490047
CAS Number: 1824436-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR