(4-Chloro-2-fluoro-3-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53125-500MG 53125-1G 53125-5G
77335-600EA 360.5 USD
77335-600 77352-642 77376-882
(4-Chloro-2-fluoro-3-iodophenyl)(methyl)sulfane ≥97%
(4-Chloro-2-fluoro-3-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅ClFIS
MW: 302.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756977
CAS Number: 2705601-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR