6-Chloro-2-methoxy-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53118-500MG 53118-1G 53118-5G 53118-25G
77328-926EA 120.51 USD
77328-926 77343-744 77365-220 77376-878
6-Chloro-2-methoxy-3-(methylthio)benzaldehyde ≥97%
6-Chloro-2-methoxy-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₂S
MW: 216.685 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756975
CAS Number: 2918917-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR