4-Fluoro-2-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-2-methylthiophenol, 4-Fluoro-2-(methylthio)phenol

53104-500MG 53104-1G 53104-5G 53104-25G
77328-924EA 193.64 USD
77328-924 77352-640 77365-216 77387-290
4-Fluoro-2-(methylthio)phenol ≥97%
4-Fluoro-2-methylthiophenol

Precaución: for Research Use Only

Formula: C₇H₇FOS
Storage Temperature: Ambient
MDL Number: MFCD16998914
CAS Number: 90033-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR