1-(3,5-Di-tert-butylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

53098-500MG 53098-1G 53098-5G 53098-25G
77328-922EA 255.44 USD
77328-922 77352-638 77365-214 77387-288
1-(3,5-Di-tert-butylphenyl)ethanone ≥95%
1-(3,5-Di-tert-butylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₆H₂₄O
Storage Temperature: Ambient
MDL Number: MFCD00598157
CAS Number: 1756-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR