4-(Benzyloxy)-1-fluoro-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

53095-500MG 53095-1G 53095-5G 53095-25G
77328-920EA 100.94 USD
77328-920 77342-928 77365-212 77376-874
4-(Benzyloxy)-1-fluoro-2-iodobenzene ≥97%
4-(Benzyloxy)-1-fluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀FIO
MW: 328.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29040512
CAS Number: 346425-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR