1-(5-(Benzyloxy)-2-fluorophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

53093-500MG 53093-1G 53093-5G 53093-25G
77328-918EA 225.57 USD
77328-918 77352-636 77365-210 77387-286
1-(5-(Benzyloxy)-2-fluorophenyl)ethanone ≥97%
1-(5-(Benzyloxy)-2-fluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
MW: 244.261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29040552
CAS Number: 1558246-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR