(2-Chloro-3-ethoxy-6-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53090-500MG 53090-1G 53090-5G 53090-25G
77328-912EA 141.11 USD
77328-912 77344-828 77365-204 77387-284
(2-Chloro-3-ethoxy-6-fluorophenyl)(methyl)sulfane ≥97%
(2-Chloro-3-ethoxy-6-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀ClFOS
MW: 220.692 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178662
CAS Number: 2643368-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR