(2-Fluoro-4,5-dimethoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53088-500MG 53088-1G 53088-5G 53088-25G
77328-910EA 120.51 USD
77328-910 77343-742 77365-202 77376-868
(2-Fluoro-4,5-dimethoxyphenyl)(methyl)sulfane ≥97%
(2-Fluoro-4,5-dimethoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂S
Storage Temperature: Ambient
MDL Number: MFCD28793160
CAS Number: 1806314-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR