(3-Bromo-2,4,6-trimethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

53074-500MG 53074-1G 53074-5G
77328-906EA 340.93 USD
77328-906 77352-632 77376-864
(3-Bromo-2,4,6-trimethylphenyl)methanol ≥97%
(3-Bromo-2,4,6-trimethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD34178658
CAS Number: 1535410-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR