(4-(Benzylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

53060-500MG 53060-1G 53060-5G 53060-25G
77328-904EA 263.68 USD
77328-904 77352-628 77365-194 77387-280
(4-(Benzylthio)phenyl)methanol ≥97%
(4-(Benzylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD18890899
CAS Number: 3448-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR