3-(Trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

53055-500MG 53055-1G 53055-25G
77328-902EA 262.65 USD
77328-902 77352-626 77387-278
3-(Trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3-(Trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₉F₃O
MW: 250.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14702013
CAS Number: 1214364-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR