Methyl 3-(trifluoromethyl)-[1,1'-biphenyl]-4-carboxylate ≥95%

Supplier: AOBChem USA

Warning

53054-500MG 53054-1G 53054-5G
77339-758EA 628.3 USD
77339-758 77352-624 77376-858
Methyl 3-(trifluoromethyl)-[1,1'-biphenyl]-4-carboxylate ≥95%
Methyl 3-(trifluoromethyl)-[1,1'-biphenyl]-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃O₂
MW: 280.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756973
CAS Number: 957207-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR