Methyl 4-(methylthio)-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

53050-500MG 53050-1G 53050-5G
77337-276EA 410.97 USD
77337-276 77352-622 77376-856
Methyl 4-(methylthio)-3-(trifluoromethyl)benzoate ≥97%
Methyl 4-(methylthio)-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂S
MW: 250.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529303
CAS Number: 1005206-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR