4-Bromo-2-fluoro-3-iodophenol ≥97%

Supplier: AOBChem USA

Warning

53042-500MG 53042-1G 53042-5G
77328-894EA 345.05 USD
77328-894 77352-614 77376-852
4-Bromo-2-fluoro-3-iodophenol ≥97%
4-Bromo-2-fluoro-3-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃BrFIO
MW: 316.894 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28739590
CAS Number: 1805937-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR