3-Bromo-5-fluoro-2-iodobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53037-500MG 53037-1G 53037-5G
77339-072EA 548.99 USD
77339-072 77352-612 77376-850
3-Bromo-5-fluoro-2-iodobenzoic acid ≥97%
3-Bromo-5-fluoro-2-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrFIO₂
Storage Temperature: Ambient
MDL Number: MFCD34564227
CAS Number: 2092613-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR