Methyl 4-isobutylbenzoate ≥97%

Supplier: AOBChem USA

Warning

53034-500MG 53034-1G 53034-5G 53034-25G
77328-892EA 126.69 USD
77328-892 77344-130 77365-186 77376-848
Methyl 4-isobutylbenzoate ≥97%
Methyl 4-isobutylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MW: 192.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16878419
CAS Number: 154320-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR