2-Isobutoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53030-500MG 53030-1G 53030-5G 53030-25G
77328-890EA 251.32 USD
77328-890 77352-610 77365-184 77387-270
2-Isobutoxy-5-methylbenzoic acid ≥97%
2-Isobutoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11901722
CAS Number: 105475-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR