2-Isobutoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53028-500MG 53028-1G 53028-5G 53028-25G
77328-888EA 141.11 USD
77328-888 77344-826 77365-182 77387-268
2-Isobutoxy-5-methylbenzaldehyde ≥97%
2-Isobutoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD18331322
CAS Number: 1340411-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR