2-Iodo-1-isobutoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

53027-500MG 53027-1G 53027-5G
77328-886EA 339.9 USD
77328-886 77352-608 77376-846
2-Iodo-1-isobutoxy-4-methylbenzene ≥97%
2-Iodo-1-isobutoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅IO
Storage Temperature: Ambient
MDL Number: MFCD32176629
CAS Number: 1864010-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR