2,6-Dibromo-4-isobutylphenol ≥97%

Supplier: AOBChem USA

Warning

53024-500MG 53024-1G 53024-5G 53024-25G
77328-884EA 223.51 USD
77328-884 77352-606 77365-180 77387-266
2,6-Dibromo-4-isobutylphenol ≥97%
2,6-Dibromo-4-isobutylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₂Br₂O
Storage Temperature: Ambient
MDL Number: MFCD34178654
CAS Number: 2643367-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR