3',4'-Difluoro-2,5-dimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

53018-500MG 53018-1G 53018-5G
77328-876EA 346.08 USD
77328-876 77352-598 77376-840
3',4'-Difluoro-2,5-dimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3',4'-Difluoro-2,5-dimethoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₂O₃
MW: 278.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404113
CAS Number: 2635937-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR