4'-Chloro-2,5-dimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

53016-500MG 53016-1G 53016-5G
77328-872EA 346.08 USD
77328-872 77352-594 77376-836
4'-Chloro-2,5-dimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'-Chloro-2,5-dimethoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₃
Storage Temperature: Ambient
MDL Number: MFCD33404111
CAS Number: 2635937-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR