2,5-Dimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

53014-500MG 53014-1G 53014-5G
77328-870EA 307.97 USD
77328-870 77352-592 77376-834
2,5-Dimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2,5-Dimethoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404110
CAS Number: 109074-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR