5-Chloro-2-methyl-4-(pyridin-4-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53008-500MG 53008-1G 53008-5G
77328-868EA 319.3 USD
77328-868 77352-586 77376-828
5-Chloro-2-methyl-4-(pyridin-4-yl)benzaldehyde ≥97%
5-Chloro-2-methyl-4-(pyridin-4-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClNO
MW: 231.678 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530189
CAS Number: 2734772-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR