1-Bromo-4-ethyl-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52993-500MG 52993-1G 52993-5G
77338-660EA 507.79 USD
77338-660 77352-582 77376-824
1-Bromo-4-ethyl-2-methylbenzene ≥97%
1-Bromo-4-ethyl-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁Br
Storage Temperature: Ambient
CAS Number: 90560-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR