Methyl 3-bromo-5-(tert-butyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

52984-500MG 52984-1G 52984-5G 52984-25G
77328-864EA 184.37 USD
77328-864 77347-220 77365-174 77387-258
Methyl 3-bromo-5-(tert-butyl)benzoate ≥95%
Methyl 3-bromo-5-(tert-butyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrO₂
MW: 271.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840763
CAS Number: 560131-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR