3,5-Dibromo-2,6-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

52971-500MG 52971-1G 52971-5G
77335-684EA 363.59 USD
77335-684 77352-574 77376-820
3,5-Dibromo-2,6-difluorophenol ≥97%
3,5-Dibromo-2,6-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₂Br₂F₂O
MW: 287.884 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28784362
CAS Number: 1803783-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR