1,5-Dibromo-2,4-difluoro-3-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

52970-1G 52970-5G 52970-25G
77341-600EA 94.76 USD
77341-600 77365-168 77376-818
1,5-Dibromo-2,4-difluoro-3-iodobenzene ≥95%
1,5-Dibromo-2,4-difluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆HBr₂F₂I
Storage Temperature: Ambient
MDL Number: MFCD28783628
CAS Number: 679836-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR