1,5-Dibromo-2,4-difluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52968-500MG 52968-1G 52968-5G
77337-560EA 422.3 USD
77337-560 77352-572 77376-816
1,5-Dibromo-2,4-difluoro-3-methylbenzene ≥97%
1,5-Dibromo-2,4-difluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₄Br₂F₂
Storage Temperature: Ambient
MDL Number: MFCD28784486
CAS Number: 1806351-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR