5-Chloro-[1,1'-biphenyl]-3-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

52942-500MG 52942-1G 52942-5G
77335-554EA 357.41 USD
77335-554 77352-568 77376-812
5-Chloro-[1,1'-biphenyl]-3-carbaldehyde ≥95%
5-Chloro-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉ClO
Storage Temperature: Ambient
MDL Number: MFCD34178652
CAS Number: 1356967-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR