Methyl 5-chloro-[1,1'-biphenyl]-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

52940-500MG 52940-1G 52940-5G 52940-25G
77328-854EA 259.56 USD
77328-854 77352-564 77365-164 77387-250
Methyl 5-chloro-[1,1'-biphenyl]-3-carboxylate ≥97%
Methyl 5-chloro-[1,1'-biphenyl]-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₂
MW: 246.689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27578693
CAS Number: 1820703-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR