(5-Bromo-2,4-dimethylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

52938-1G 52938-5G 52938-25G
77341-446EA 86.52 USD
77341-446 77365-162 77376-808
(5-Bromo-2,4-dimethylphenyl)(methyl)sulfane ≥95%
(5-Bromo-2,4-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁BrS
MW: 231.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178651
CAS Number: 2167757-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR