(5-Bromo-2,4-dimethylphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

52934-500MG 52934-1G 52934-25G 52934-5G
77328-848EA 305.91 USD
77328-848 77352-558 77387-244 77376-800
(5-Bromo-2,4-dimethylphenyl)(morpholino)methanone ≥97%
(5-Bromo-2,4-dimethylphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrNO₂
MW: 298.176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178648
CAS Number: 2643368-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR