(5-Bromo-2,4-dimethylphenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

52933-500MG 52933-1G 52933-25G 52933-5G
77328-846EA 305.91 USD
77328-846 77352-556 77387-242 77376-798
(5-Bromo-2,4-dimethylphenyl)(piperidin-1-yl)methanone ≥97%
(5-Bromo-2,4-dimethylphenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO
MW: 296.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530187
CAS Number: 2734776-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR