(5-Bromo-2,4-dimethylphenyl)(pyrrolidin-1-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

52932-500MG 52932-1G 52932-5G
77328-844EA 326.51 USD
77328-844 77352-554 77376-796
(5-Bromo-2,4-dimethylphenyl)(pyrrolidin-1-yl)methanone ≥95%
(5-Bromo-2,4-dimethylphenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrNO
MW: 282.176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530186
CAS Number: 2734776-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR