5-Bromo-N,N,2,4-tetramethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

52929-500MG 52929-1G 52929-25G 52929-5G
77328-838EA 305.91 USD
77328-838 77352-548 77387-236 77376-790
5-Bromo-N,N,2,4-tetramethylbenzamide ≥97%
5-Bromo-N,N,2,4-tetramethylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO
MW: 256.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530184
CAS Number: 2734772-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR