1-(Benzyloxy)-5-bromo-2,4-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

52924-500MG 52924-1G 52924-5G
77338-706EA 510.88 USD
77338-706 77352-546 77376-786
1-(Benzyloxy)-5-bromo-2,4-dimethylbenzene ≥97%
1-(Benzyloxy)-5-bromo-2,4-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO
MW: 291.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178644
CAS Number: 2643368-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR