5-Iodo-2,4-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52912-500MG 52912-1G 52912-5G
77338-142EA 466.59 USD
77338-142 77352-544 77376-784
5-Iodo-2,4-dimethylbenzaldehyde ≥97%
5-Iodo-2,4-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉IO
Storage Temperature: Ambient
MDL Number: MFCD34530183
CAS Number: 1026162-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR