3-Ethoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52907-500MG 52907-1G 52907-5G
77328-834EA 347.11 USD
77328-834 77352-542 77376-782
3-Ethoxy-5-methylbenzaldehyde ≥97%
3-Ethoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34178641
CAS Number: 2643368-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR