3-Isopropoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52901-500MG 52901-1G 52901-5G
77338-972EA 538.69 USD
77338-972 77352-540 77376-780
3-Isopropoxy-5-methylbenzaldehyde ≥97%
3-Isopropoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD29122313
CAS Number: 1289090-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR