Methyl 3-(benzyloxy)-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

52892-500MG 52892-1G 52892-5G 52892-25G
77328-832EA 184.37 USD
77328-832 77347-218 77365-156 77387-234
Methyl 3-(benzyloxy)-5-methylbenzoate ≥97%
Methyl 3-(benzyloxy)-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MW: 256.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34702812
CAS Number: 189252-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR